4-Fluoro-2-nitrotoluene ≥98%

Warning

AC11952-0100 AC11952-0250
200035-884EA 66.5 USD
200035-884 200035-882
4-Fluoro-2-nitrotoluene ≥98%
4-Fluoro-2-nitrotoluene
Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 27 °C
Density: 1,2911 g/cm³ (0 °C)
MDL Number: MFCD00007200
CAS Number: 446-10-6
EINECS: 207-161-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear yellow to amber Liquid
Infrared spectrum Conforms
Refractive index 1.5218 to 1.5238 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR