2-Fluorobenzoic acid 99%

Warning

Synonyms: o-Fluorobenzoic acid

AC11936-1000 AC11936-0250
200016-560EA 36.9 USD
200016-560 200016-562
2-Fluorobenzoic acid 99%
2-Fluorobenzoic acid
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 123…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or needles
Infrared spectrum Conforms
Melting point 122 to 125 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR