4-Fluoroanisole 99%

Warning

Synonyms: 1-Fluoro-4-methoxybenzene

AC11931-0250 AC11931-1000
200017-526EA 238 USD
200017-526 200029-822
4-Fluoroanisole 99%
4-Fluoroanisole
Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 157 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.11 g/cm³
Flash Pt: 43 °C
MDL Number: MFCD00000348
CAS Number: 459-60-9
EINECS: 207-295-7
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4867 to 1.4887 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR