4-Fluoroaniline 98%

Danger

AC11929-5000 AC11929-1000 AC11929-0250
200010-410EA 171 USD
200010-410 200010-408 103524-044
4-Fluoroaniline 98%
4-Fluoroaniline
Formula: C₆H₆FN
MW: 111.12 g/mol
Boiling Pt: 187 °C (760 mmHg)
Melting Pt: –1.9 °C
Density: 1.157 g/cm³
Flash Pt: 73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007829
CAS Number: 371-40-4
EINECS: 206-735-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,04196

Order Now

Specification Test Results

Appearance Clear light yellow to red-brown Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5385 to 1.5405 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR