9-Fluorenone-2-carboxylic acid 97%

Synonyms: 9-Oxofluoren-2-carboxylic acid

AC11921-0010
200036-422EA 30.5 USD
200036-422
9-Fluorenone-2-carboxylic acid 97%
9-Fluorenone-2-carboxylic acid
Formula: C₁₄H₈O₃
MW: 224.22 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: >300 °C
MDL Number: MFCD00001156
CAS Number: 784-50-9
EINECS: 212-317-3

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
GC ≥96.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR