Ethyl propiolate 99%

Warning

Synonyms: Propiolic acid ethyl esterEthyl acetylenecarboxylate2-Propynoic acid ethyl esterEthyl propynoate

AC11890-1000 AC11890-0050 AC11890-0250
103525-080EA 342 USD
103525-080 200006-506 200006-504
Ethyl propiolate 99%
Ethyl propiolate
Formula: C₅H₆O₂
MW: 98.1 g/mol
Boiling Pt: 120 °C (760 mmHg)
Density: 0.968 g/cm³
Flash Pt: 23 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009184
CAS Number: 623-47-2
EINECS: 210-795-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4110 to 1.4130 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR