4-Ethylphenol 97%

Danger

AC11882-2500 AC11882-0010 AC11882-0051 AC11882-0050
200035-870EA 182 USD
200035-870 103530-646 200010-398 200041-990
4-Ethylphenol 97%
4-Ethylphenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 218…219 °C (760 mmHg)
Melting Pt: 41…46 °C
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002393
CAS Number: 123-07-9
EINECS: 204-598-6
UN: 2430
ADR: 8,III

Order Now

Specification Test Results

Appearance White to cream to brown Crystals or crystalline solid or fused mass
Infrared spectrum Conforms
Melting point 38 to 45 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR