Ethylene carbonate 99+%

Warning

Synonyms: 1,3-Dioxa-2-cyclopentanone

AC11841-0010 AC11841-2500
200000-452EA 48.4 USD
200000-452 200012-466
Ethylene carbonate 99+%
Ethylene carbonate
Formula: C₃H₄O₃
MW: 88.06 g/mol
Boiling Pt: 246.7 °C (760 mmHg)
Melting Pt: 34…37 °C
Density: 1.321 g/cm³
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00005382
CAS Number: 96-49-1
EINECS: 202-510-0

Order Now

Specification Test Results

Appearance White to yellow Low melting crystalline solid
Infrared spectrum Conforms
Melting point 34 to 37 °C
GC ≥99.0 %
Specific optical rotation +66.3° to +66.8° (25 °C, 589 nm) (c=26, H2O)
Chloride (Cl) ≤50 ppm
SO4 and SO3 (as SO4) ≤0.005 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm
Titrable acid ≤0.0008 meq/g
Insoluble matter ≤0.005 %
Invert sugar ≤0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR