Ethyl cyanoacetate 98+%

Warning

AC11828-0010 AC11828-2500
200000-450EA 45.6 USD
200000-450 200012-404
Ethyl cyanoacetate 98+%
Ethyl cyanoacetate
Formula: C₅H₇NO₂
MW: 113.12 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: –22 °C
Density: 1.06 g/cm³
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001940
CAS Number: 105-56-6
EINECS: 203-309-0
Merck Index: 13,03821

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20 °C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.0 %
Color scale ≤50 APHA
Free acid ≤10 mg/kg (as cyanoacetic acid)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR