N-Ethylaniline ≥98%

Supplier: ACROS ORGANICS MS

Danger

Synonyms: N-Ethyl-N-phenylamine

AC11804-5000 AC11804-1000 AC11804-0025
200044-168EA 50.29 USD
200044-168 200020-046 200038-178
N-Ethylaniline ≥98%
N-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 204,7 °C (1013 hPa)
Melting Pt: –64 °C
Density: 0.96 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00009025
CAS Number: 103-69-5
EINECS: 203-135-5
UN: 2272
ADR: 6.1,III
Merck Index: 13,03800

Order Now

Specification Test Results

Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5530 to 1.5560 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR