Ethyl-3-aminobenzoate 99+%

Warning

Synonyms: Ethyl 3-aminobenzoate

AC11799-0050 AC11799-1000 AC11799-0250
200043-500EA 33 USD
200043-500 200025-366 103509-462
Ethyl-3-aminobenzoate 99+%
Ethyl 3-aminobenzoate
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 172…175 °C (13 mmHg)
Density: 1.107 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00007794
CAS Number: 582-33-2
EINECS: 209-482-9

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid or low melting solid
Infrared spectrum Conforms
Refractive index 1.5595 to 1.5615 (20 °C, 589 nm)
GC ≥99.0 %
Chloride (Cl) ≤10 ppm
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.02 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR