Diethylene glycol monoethyl ether 98+%

Synonyms: 2-(2-Ethoxyethoxy)ethanolDi(ethylene glycol) ethyl etherDiethylene Glycol Monoethyl Ether

AC11789-0010 AC11789-0025 AC11789-0250
200004-820EA 160.38 USD
200004-820 103465-860 200004-818
Diethylene glycol monoethyl ether 98+%
Diethylene glycol monoethyl ether
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 197 °C (760 mmHg)
Melting Pt: –80 °C
Density: 0.99 g/cm³
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 13,01809

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4280 (20 °C, 589 nm)
GC ≥98.0 %
Sulfate (SO4) ≤0.003 %
Potassium carbonate ≤2.0 % (K2CO3)
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.002 %
Nickel (Ni) ≤0.001 %
Sodium (Na) ≤0.05 %
Heavy metals (ICP-OES) ≤0.001 %
Nitrogen compounds (as N) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR