1,2-Ethanedithiol 95%

Danger

Synonyms: 1,2-Dimercaptoethane1.2-Ethanedithiol

AC11785-0010
103525-030EA 304 USD
103525-030
1,2-Ethanedithiol 95%
1,2-Ethanedithiol
Formula: C₂H₆S₂
MW: 94.2 g/mol
Boiling Pt: 144…146 °C (760 mmHg)
Melting Pt: –41 °C
Density: 1.123 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00004892
CAS Number: 540-63-6
EINECS: 208-752-3
UN: 3071
ADR: 6.1,II
Merck Index: 13,03760

Order Now

Specification Test Results

Appearance Clear colorless to yellow to slightly colored Liquid
Infrared spectrum Conforms
GC ≥94 %
Bromide (Br) ≤0.01 %
Iodide (I) ≤20 ppm
Chlorate and Nitrate (as NO3) ≤0.003 %
Phosphate (PO4) ≤5 ppm
Sulfate (SO4) ≤10 ppm
Barium (Ba) Passes test (Limit about 0.001%)
Calcium (Ca) ≤20 ppm
Iron (Fe) ≤3 ppm
Magnesium (Mg) ≤10 ppm
Sodium (Na) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR