1-Dodecene 93-95%

Danger

Synonyms: Dodec-1-ene

AC11763-5000 AC11763-1000
200015-786EA 48.1 USD
200015-786 200021-818
1-Dodecene 93-95%
1-Dodecene
Formula: C₁₂H₂₄
MW: 168.32 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: –35 °C
Density: 0.75 g/cm³
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00008961
CAS Number: 112-41-4
EINECS: 203-968-4
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4284 to 1.4304 (20 °C, 589 nm)
GC ≥93 %
Solubility (In water) Passes test
GC ≥99.8 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Substance darkened by H2SO4 Passes test
Carbonyl compounds ≤0.001 %
Carbonyl compounds Each of acetone, formaldehyde and acetaldehyde
Titrable acid ≤0.0003 meq/g
Titrable base ≤0.0002 meq/g
KMnO4-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR