1-Dodecanethiol 98%

Danger

Synonyms: n-Dodecyl mercaptan1-Mercaptododecanen-Dodecanethiol

AC11762-0025 AC11762-0100 AC11762-5000
103525-024EA 101 USD
103525-024 200000-432 103525-026
1-Dodecanethiol 98%
1-Dodecanethiol
Formula: C₁₂H₂₆S
MW: 202.4 g/mol
Boiling Pt: 266…283 °C (760 mmHg)
Melting Pt: –7 °C
Density: 0.84 g/cm³
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00004885
CAS Number: 112-55-0
EINECS: 203-984-1
UN: 1760
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4580 to 1.4600 (20 °C, 589 nm)
GC ≥97.5 %
Color scale ≤20 APHA
Residue after reduction ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR