n-Dodecane ≥99%, pure

Supplier: ACROS ORGANICS MS

Danger

AC11759-2500 AC11759-0250 AC11759-1000 AC11759-0025 AC11759-0100 AC11759-5000
103466-122EA 104.29 USD InStock
103466-122 200012-264 200000-428 103466-118 103466-120 200000-430
n-Dodecane ≥99%, pure
n-Dodecane
Formula: H₃C(CH₂)₁₀CH₃
MW: 170.34 g/mol
Boiling Pt: 215°C - 217°C (760mmHg)
Melting Pt: –12 °C
Density: 0.7487 g/cm³ (20 °C)
Flash Pt: 71°C
Storage Temperature: Ambient
MDL Number: MFCD00008969
CAS Number: 112-40-3
EINECS: 203-967-9
https://us.vwr.com/stibo/web/std.lang.all/61/61/736161.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4210 to 1.4230 (20°C, 589 nm)
GC ≥98.5 %
Color scale ≤10 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR