1,1-Diphenylethylene 98%

Synonyms: asym. DiphenyletheneEthene-1,1-diyldibenzeneasym. Diphenylethylene

AC11726-0100 AC11726-0250 AC11726-1000
103509-296EA 421 USD
103509-296 200008-386 200008-384
1,1-Diphenylethylene 98%
1,1-Diphenylethylene
Formula: (C₆H₅)₂CCH₂
MW: 180.25 g/mol
Boiling Pt: 270…271 °C (760 mmHg)
Melting Pt: 6 °C
Density: 1.02 g/cm³
Flash Pt: >112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008583
CAS Number: 530-48-3
EINECS: 208-482-6
Merck Index: 13,03355
https://us.vwr.com/stibo/web/std.lang.all/07/18/760718.jpg

Order Now

Specification Test Results

Appearance Clear colorless to golden Liquid
Infrared spectrum Conforms
Refractive index 1.6070 to 1.6090 (20 °C, 589 nm)
GC ≥97.5 %
Iodide (I) ≤30 ppm
Phosphate (PO4) ≤10 ppm
SO4 and SO3 (as SO4) ≤0.003 %
Iron (Fe) ≤1 ppm
Selenium (Se) ≤0.01 ppm
Heavy metals (ICP-OES) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR