Diphenylacetic acid 99+%

Warning

Synonyms: 2,2-Diphenylacetic acid

AC11715-5000 AC11715-1000 AC11715-0050
200049-122EA 410 USD
200049-122 200006-358 200012-150
Diphenylacetic acid 99+%
Diphenylacetic acid
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00004251
CAS Number: 117-34-0
EINECS: 204-185-0
Merck Index: 13,03348

Order Now

Specification Test Results

Appearance White to off-white to cream Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 146 to 149 °C
Titration with NaOH ≥99.0 %
Calcium (Ca) ≤0.01 %
Copper (Cu) ≤0.003 %
Magnesium (Mg) ≤0.005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.02 %
Zinc (Zn) ≤0.003 %
Ferrous iron (Fe 2+) (Limit about 0.001%) Passes test
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR