2,4-Dinitroaniline 99%

Danger

AC11693-1000 AC11693-0050 AC11693-5000
200028-288EA 286 USD
200028-288 103530-584 103530-582
2,4-Dinitroaniline 99%
2,4-Dinitroaniline
Formula: C₆H₅N₃O₄
MW: 183.12 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 177…180 °C
Flash Pt: 224 °C
Storage Temperature: Ambient
MDL Number: MFCD00007151
CAS Number: 97-02-9
EINECS: 202-553-5
UN: 1596
ADR: 6.1,II
Merck Index: 13,03298

Order Now

Specification Test Results

Appearance Yellow to yellow-green to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 176 to 180 °C
Water 8 to 15 % (K.F.)
HPLC ≥98.5 %
Copper (Cu) ≤20 ppm
Iron (Fe) ≤50 ppm
Magnesium (Mg) ≤50 ppm
Nickel (Ni) ≤1000 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤500 ppm
Zinc (Zn) ≤300 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR