2,5-Xylenol 99+%

Danger

Synonyms: 2,5-Dimethylphenol2-Hydroxy-4-xylene2-Hydroxy-p-xylene

AC11654-0050 AC11654-5000 AC11654-1000
200016-174EA 50 USD
200016-174 200046-484 200021-940
2,5-Xylenol 99+%
2,5-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 212 °C (1013 hPa)
Melting Pt: 75…77 °C
Density: 0.97 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00002237
CAS Number: 95-87-4
EINECS: 202-461-5
UN: 2261
ADR: 6.1,II
Merck Index: 12,10215

Order Now

Specification Test Results

Appearance White to cream Crystals
Infrared spectrum Conforms
Melting point ≥75 °C
Water ≤0.5 %
Sulfated ash ≤0.2 %
GC ≥99.0 %
Phosphate (PO4) ≤5 ppm
Iron (Fe) ≤5 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR