Dimethyl carbonate 99%

Danger

AC11612-5000 AC11612-0010 AC11612-0025 AC11612-0100
103526-964EA 214 USD
103526-964 200006-318 200063-346 200025-050
Dimethyl carbonate 99%
Dimethyl carbonate
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 90 °C (760 mmHg)
Melting Pt: 2…4 °C
Density: 1.069 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008420
CAS Number: 616-38-6
EINECS: 210-478-4
UN: 1161
ADR: 3,II
Merck Index: 12,06117

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.3672 to 1.3692 (20 °C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥98.5 %
Color scale ≤30 APHA
Acidity (HCl) ≤0.1 %
Methanol ≤0.2 % (GC)
Iron (Fe) ≤0.2 ppm
Heavy metals (as Pb) ≤0.5 ppm
Titrable base ≤0.0004 meq/g
KMnO4-reducing substance Passes test
K2Cr2O7-reducing substance Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR