4,4'-Dimethylbiphenyl

Synonyms: 4,4'-Bitolyl4,4'-Dimethyldiphenyl

AC11608-0010
200047-668EA 37.3 USD
200047-668
4,4'-Dimethylbiphenyl
4,4'-Dimethylbiphenyl
Formula: C₁₄H₁₄
MW: 182.27 g/mol
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 117…122 °C
Density: 1.102 g/cm³ (20 °C)
MDL Number: MFCD00008545
CAS Number: 613-33-2
EINECS: 210-337-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 117 to 122 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR