3,5-Dimethylbenzoic acid 99%

Warning

AC11600-1000 AC11600-0050 AC11600-0250
200023-096EA 235 USD
200023-096 200023-094 103465-158
3,5-Dimethylbenzoic acid 99%
3,5-Dimethylbenzoic acid
Formula: C₉H₁₀O₂
MW: 150.18 g/mol
Melting Pt: 169…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00002525
CAS Number: 499-06-9
EINECS: 207-876-5

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 169 to 172 °C
Titration with NaOH ≥98.5 %
Substance darkened by H2SO4 Passes test
Chloride (Cl) ≤0.002 %
Sulfate (SO4) ≤0.005 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤2 ppm
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR