3,5-Dimethylanisole 99%

Synonyms: 1-Methoxy-3,5-dimethylbenzene

AC11596-0250 AC11596-1000
200009-734EA 70.5 USD
200009-734 103465-156
3,5-Dimethylanisole 99%
3,5-Dimethylanisole
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 193 °C (760 mmHg)
Density: 0.96 g/cm³
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008398
CAS Number: 874-63-5
EINECS: 212-865-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5110 to 1.5130 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR