2,3-Dimethylaniline ≥99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-Amino-o-xylene
, 2,3-Xylidine
11593-0050
TS11593-0050EA
28
USD
TS11593-0050
2,3-Dimethylaniline ≥99%
2,3-Dimethylaniline
Formula:
C₈H₁₁N MW: 121.18 g/mol Boiling Pt: 222 °C (1013 hPa) Melting Pt: –15 °C Density: 0.99 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007732 CAS Number: 87-59-2 EINECS: 201-755-0 UN: 1711 ADR: 6.1,II Merck Index: 12,10217 |
Specification Test Results
Appearance | Clear yellow to red-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5670 to 1.5690 (20°C, 589 nm) |
GC | ≥98.5 % |
Titration with HClO4 | 98.5 to 101.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...