1-(Dimethylamino)-2-propanol 99%

Danger

Synonyms: DimepranolN,N-DimethylisopropanolamineN,N-Dimethyl-1-amino-2-propanol

AC11587-0010 AC11587-2500
200027-860EA 38.3 USD
200027-860 200015-780
1-(Dimethylamino)-2-propanol 99%
1-(Dimethylamino)-2-propanol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 125…127 °C (1013 hPa)
Melting Pt: –85 °C
Density: 0.85 g/cm³
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00004532
CAS Number: 108-16-7
EINECS: 203-556-4
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May discolor to yellow on storage)
Infrared spectrum Conforms
Refractive index 1.4183 to 1.4203 (20 °C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR