2,6-Dimethoxyphenol 99%

Warning

AC11559-5000 AC11559-0250 AC11559-1000
103509-254EA 513 USD
103509-254 200009-074 200016-214
2,6-Dimethoxyphenol 99%
2,6-Dimethoxyphenol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 261 °C (760 mmHg)
Melting Pt: 50…56 °C
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00064434
CAS Number: 91-10-1
EINECS: 202-041-1

Order Now

Specification Test Results

Appearance White to light yellow to grey to brown Crystalline powder or crystals or flakes or fused solid
Infrared spectrum Conforms
Melting point 52 to 57 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR