3,4-Dimethoxycinnamic acid predominantly trans 99%

Warning

Synonyms: 3,4-Dimethoxycinnamic Acid

AC11551-1000 AC11551-0050 AC11551-0250
200035-158EA 141 USD
200035-158 200035-156 200029-086
3,4-Dimethoxycinnamic acid predominantly trans 99%
3,4-Dimethoxycinnamic acid
Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 181…184 °C
MDL Number: MFCD00004387
CAS Number: 2316-26-9
EINECS: 219-025-5

Order Now

Specification Test Results

Appearance Light yellow to light beige Powder
Infrared spectrum Conforms
Melting point 181 to 184 °C
GC ≥98.5 %
Titration with NaOH ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR