3,4-Dimethoxybenzyl alcohol 96%

Warning

Synonyms: (3,4-Dimethoxyphenyl)methanolVeratryl alcohol

AC11549-5000 AC11549-1000 AC11549-0250
200016-864EA 49.7 USD
200016-864 200016-866 103509-224
3,4-Dimethoxybenzyl alcohol 96%
3,4-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 296…297 °C (732 mmHg)
Melting Pt: 22 °C
Density: 1.15 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004638
CAS Number: 93-03-8
EINECS: 202-212-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow to golden Viscous liquid
Infrared spectrum Conforms
GC ≥95 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR