3,4-Dimethoxybenzoic acid 99+%

Warning

Synonyms: Veratric acid

AC11545-1000 AC11545-0250
200016-862EA 59.5 USD
200016-862 103509-220
3,4-Dimethoxybenzoic acid 99+%
3,4-Dimethoxybenzoic acid
Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Melting Pt: 179…182 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002500
CAS Number: 93-07-2
EINECS: 202-215-7
Merck Index: 12,10086

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 180 to 184 °C
HPLC ≥99.0 %
Color scale ≤10 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR