3,5-Dimethoxyaniline 98%

Warning

AC11536-1000 AC11536-0250 AC11536-0050
200016-894EA 44.7 USD
200016-894 200009-716 200009-718
3,5-Dimethoxyaniline 98%
3,5-Dimethoxyaniline
Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
Boiling Pt: 178 °C (20 mmHg)
Melting Pt: 53…55 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00008392
CAS Number: 10272-07-8
EINECS: 233-616-5
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige to grey to light brown Powder or crystals or needles
Infrared spectrum Conforms
Melting point 54 to 57 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR