Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,8-Dihydroxy-9,10-anthraquinone

Supplier: ACROS ORGANICS MS

Synonyms: Dantron1,8-Dihydroxyanthraquinone

AC11483-5000 AC11483-0050 AC11483-1000
200045-848EA 156.15 USD InStock
200045-848 200015-638 200045-846
1,8-Dihydroxy-9,10-anthraquinone
1,8-Dihydroxy-9,10-anthraquinone


Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Melting Pt: 190…193 °C
Density: 1.537 g/cm³ (20 °C)
Flash Pt: >200 °C
MDL Number: MFCD00001211
CAS Number: 117-10-2
EINECS: 204-173-5
Merck Index: 13,02841
Order Now
Specification Test Results
Appearance Yellow-green to orange to brown Powder
Infrared spectrum Conforms
Melting point 190°C to 195°C
HPLC ≥94 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty