Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Difluoroaniline ≥99%

Supplier: ACROS ORGANICS MS

Warning

AC11459-0100 AC11459-0250 AC11459-1000
200016-102EA 61.07 USD InStock
200016-102 200034-354 103530-550
2,4-Difluoroaniline ≥99%
2,4-Difluoroaniline


Formula: C₆H₅F₂N
MW: 129.11 g/mol
Boiling Pt: 170 °C (753 mmHg)
Melting Pt: –7,5 °C
Density: 1.268 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00007648
CAS Number: 367-25-9
EINECS: 206-687-5
UN: 2941
ADR: 6.1,III
Merck Index: 13,03171
Order Now
Specification Test Results
Appearance Clear brown Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5025 to 1.5075 (20°C, 589 nm)
GC ≥98.5 %


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty