Diethyl phthalate 99%

Synonyms: Phthalic acid diethyl ester

AC11452-0010 AC11452-0100 AC11452-0250 AC11452-0025
103509-184EA 252 USD
103509-184 200006-284 200006-282 200000-360
Diethyl phthalate 99%
Diethyl phthalate
Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Boiling Pt: 298…299 °C (760 mmHg)
Density: 1.118 g/cm³
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00009111
CAS Number: 84-66-2
EINECS: 201-550-6
Merck Index: 12,07527
https://us.vwr.com/stibo/web/std.lang.all/99/50/2299950.jpg

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.5010 to 1.5030 (20 °C, 589 nm)
Water ≤0.2 % (K.F.)
GC ≥98.5 %
Color scale ≤25 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR