Diethyl fumarate 98%

Warning

Synonyms: Fumaric acid diethyl ester

AC11438-0010 AC11438-2500 AC11438-0100 AC11438-0025
200030-964EA 22.8 USD
200030-964 103509-100 200024-998 200018-780
Diethyl fumarate 98%
Diethyl fumarate
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 218…219 °C (760 mmHg)
Melting Pt: 1…2 °C
Density: 1.05 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00064455
CAS Number: 623-91-6
EINECS: 210-819-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4396 to 1.4416 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR