2-Diethylaminoethanol 99%

Danger

Synonyms: N,N-Diethylethanolamine

AC11433-0100
103524-940EA 243 USD
103524-940
2-Diethylaminoethanol 99%
2-Diethylaminoethanol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 161 °C (760 mmHg)
Melting Pt: –70 °C
Density: 0.88 g/cm³
Flash Pt: 51.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00002850
CAS Number: 100-37-8
EINECS: 202-845-2
UN: 2686
ADR: 8,II
Merck Index: 13,03139

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4400 to 1.4420 (20 °C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥98.5 %
Color scale ≤50 APHA

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR