2,6-Dichlorotoluene 99+%

Warning

Synonyms: 1,3-Dichloro-2-methylbenzene

AC11379-1000 AC11379-0025 AC11379-5000
200022-320EA 50.1 USD
200022-320 200028-370 103530-532
2,6-Dichlorotoluene 99+%
2,6-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 196…203 °C (760 mmHg)
Melting Pt: 2 °C
Density: 1.25 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00000576
CAS Number: 118-69-4
EINECS: 204-269-7
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5500 to 1.5520 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR