1,2-Dichloropropane 98%, pure

Danger

Synonyms: Propylene dichloride

AC11367-0050 AC11367-0010 AC11367-2500 AC11367-0500
103524-920EA 183.46 USD
103524-920 200027-578 103524-918 200027-580
1,2-Dichloropropane 98%, pure
1,2-Dichloropropane
Formula: ClH₂CCH₂CH₂Cl
MW: 112.99 g/mol
Boiling Pt: 95…96 °C (760 mmHg)
Melting Pt: –100 °C
Density: 1.156 g/cm³
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00000868
CAS Number: 78-87-5
EINECS: 201-152-2
UN: 1279
ADR: 3,II
Merck Index: 12,08039

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4400 (20 °C, 589 nm)
GC ≥97.5 %
Ash ≤4.5 % (825 °C)
pH 6.5 to 7 (10 % at 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR