Dichloromethyl methyl ether 97%

Danger

Synonyms: 1,1-Dichlorodimethyl ether

AC11347-1000 AC11347-0100
103530-516EA 142 USD
103530-516 200000-334
Dichloromethyl methyl ether 97%
Dichloromethyl methyl ether
Formula: C₂H₄Cl₂O
MW: 114.96 g/mol
Boiling Pt: 85 °C (760 mmHg)
Density: 1.27 g/cm³
Flash Pt: 22 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000828
CAS Number: 4885-02-3
EINECS: 225-498-9
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4290 to 1.4330 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR