5,7-Dichloro-8-quinolinol 98%

Warning

Synonyms: 5,7-Dichloro-8-hydroxyquinoline

AC11343-1000 AC11343-0050
200024-026EA 96.2 USD
200024-026 200036-158
5,7-Dichloro-8-quinolinol 98%
5,7-Dichloro-8-quinolinol
Formula: C₉H₅Cl₂NO
MW: 214.05 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 178…183 °C
MDL Number: MFCD00006786
CAS Number: 773-76-2
EINECS: 212-258-3
Merck Index: 13,02193

Order Now

Specification Test Results

Appearance White to beige to grey Powder
Infrared spectrum Conforms
Melting point 178 to 183 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR