Dimethyldichlorosilane 99+%

Danger

Synonyms: DichlorodimethylsilaneDichloro(dimethyl)silane

AC11331-0025
103524-910EA 158.4 USD
103524-910
Dimethyldichlorosilane 99+%
Dimethyldichlorosilane
Formula: C₂H₆Cl₂Si
MW: 129.06 g/mol
Boiling Pt: 70 °C (760 mmHg)
Melting Pt: –76 °C
Density: 1.06 g/cm³
Flash Pt: –9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000491
CAS Number: 75-78-5
EINECS: 200-901-0
UN: 1162
ADR: 3,II
REACH: 01-2119437250-51

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4030 to 1.4060 (20 °C, 589 nm)
GC ≥99.0 %
Absorption Ratio P-15/P+15 1.21 to 1.95
BSC Certification Passes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR