Dichloroacetic acid (DCA) 99+%
|
Danger
|
Synonyms:
DCA
AC11306-0025
AC11306-2500
AC11306-0010
103524-900EA
371
USD
103524-900
200000-310
200000-308
Dichloroacetic acid (DCA) 99+%
Dichloroacetic acid (DCA)
Formula:
C₂H₂Cl₂O₂ MW: 128.94 g/mol Boiling Pt: 194 °C (760 mmHg) Melting Pt: 9…11 °C Density: 1.56 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004223 CAS Number: 79-43-6 EINECS: 201-207-0 UN: 1764 ADR: 8,II Merck Index: 13,03075 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4650 to 1.4670 (20 °C, 589 nm) |
Color scale | ≤100 APHA |
Titration with NaOH | 99.0 to 101.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...