N,N-Dibutylformamide 99+%

Danger

Synonyms: FormdibutylamideFormic acid dibutylamide

AC11298-0100 AC11298-5000
103524-896EA 233.08 USD
103524-896 200044-066
N,N-Dibutylformamide 99+%
N,N-Dibutylformamide
Formula: C₉H₁₉NO
MW: 157.26 g/mol
Boiling Pt: 120 °C (15 mmHg)
Density: 0.86 g/cm³
Flash Pt: 100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003289
CAS Number: 761-65-9
EINECS: 212-090-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4430 to 1.4450 (20 °C, 589 nm)
Water ≤0.1 % (K.F.)
GC ≥99.0 %
HPLC 48.0 to 52.0 % p-Benzoquinone
HPLC 48.0 to 52.0 % Hydroquinone
Sulfate (SO4) ≤200 ppm
Iron (Fe) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR