α,α'-Dibromo-p-xylene 97%

Danger

Synonyms: p-Xylylene dibromide1,4-Bis(bromomethyl)benzene

AC11294-0250 AC11294-1000
200011-072EA 104 USD
200011-072 200018-064
α,α'-Dibromo-p-xylene 97%
α,α'-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 142…147 °C
Density: 2.012 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000182
CAS Number: 623-24-5
EINECS: 210-781-1
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or coarse crystals
Infrared spectrum Conforms
Melting point 142 to 147 °C
GC ≥96.0 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR