α,α'-Dibromo-o-xylene 96%

Danger

Synonyms: o-Xylylene bromidealpha,alpha'-Dibromo-o-xyleneo-Xylylene dibromideo-Xylene dibromide1,2-Bis(bromomethyl)benzene

AC11293-1000 AC11293-0250
200011-066EA 77.5 USD
200011-066 200011-068
α,α'-Dibromo-o-xylene 96%
α,α'-Dibromo-o-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 140 °C (20 mmHg)
Melting Pt: 91…96 °C
Density: 1.96 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000175
CAS Number: 91-13-4
EINECS: 202-042-7
UN: 2928
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 91 to 96 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR