2,6-Dibromopyridine 98%

Danger

AC11291-1000 AC11291-5000 AC11291-0250
103524-890EA 716 USD
103524-890 200009-046 200009-048
2,6-Dibromopyridine 98%
2,6-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Melting Pt: 118…119 °C
Flash Pt: 213 °C
Storage Temperature: Ambient
MDL Number: MFCD00006223
CAS Number: 626-05-1
EINECS: 210-926-9
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to grey to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 117 to 121 °C
GC ≥97.5 %
Boiling point 114 to 116 °C
GC ≥99.0 %
Color scale ≤20 APHA
Residue after evaporation ≤0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR