2,5-Dibromopyridine 97%

Warning

AC11290-0100 AC11290-0500
103530-502EA 229 USD
103530-502 200000-300
2,5-Dibromopyridine 97%
2,5-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 234…235 °C (1013 hPa)
Melting Pt: 91…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00006221
CAS Number: 624-28-2
EINECS: 210-839-6
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to beige to brown Crystalline powder
Infrared spectrum Conforms
Melting point 91 to 96 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR