3,4-Diaminopyridine 98%

Danger

Synonyms: Pyridine-3,4-diamine3,4-DAPPyridine-3,4-diyldiamine3,4-Pyridinediamine

AC11239-0250 AC11239-0050
200023-016EA 105.99 USD
200023-016 103530-496
3,4-Diaminopyridine 98%
3,4-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 216…221 °C (dec.)
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00006401
CAS Number: 54-96-6
EINECS: 200-220-9
UN: 2811
ADR: 6.1,II
Merck Index: 13,03006

Order Now

Specification Test Results

Appearance Grey-brown to brown Crystalline powder (May darken on storage)
Infrared spectrum Conforms
Melting point 216 to 221 °C (Dec)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR