2,6-Diaminopyridine 98%

Danger

AC11238-0250 AC11238-1000 AC11238-5000
103530-494EA 23.7 USD
103530-494 200009-044 200022-288
2,6-Diaminopyridine 98%
2,6-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 285 °C (760 mmHg)
Melting Pt: 115…123 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00006329
CAS Number: 141-86-6
EINECS: 205-507-2
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Beige to grey to dark brown Crystalline powder or flakes (May darken on storage)
Infrared spectrum Conforms
Melting point 115 to 123 °C
Water ≤1 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR