2,4-Diamino-6-hydroxypyrimidine 96%

Warning

Synonyms: 2,4-Diamino-6-pyrimidinol6-Aminoisocytosin2,4-Diaminopyrimidin-6-ol

AC11224-1000
200016-076EA 76.1 USD
200016-076
2,4-Diamino-6-hydroxypyrimidine 96%
2,4-Diamino-6-hydroxypyrimidine
Formula: C₄H₆N₄O
MW: 126.12 g/mol
Melting Pt: 270…290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006098
CAS Number: 56-06-4
EINECS: 200-254-4
Merck Index: 13,03001

Order Now

Specification Test Results

Appearance White to cream Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Titration with HClO4 ≥95 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR